Would you like to log in again?
OHAUS® Microplate Vortex Mixer
Supplier: Ohaus
Ohaus
30392160
470235-168EA 713 USD InStock
470235-168
OHAUS® Microplate Vortex Mixer
Shakers and Mixers Microplate Shakers
Designed for Strong, Consistent Mixing of Microplates.
  • Designed for continuous duty
  • Designed for shaking microplates or tubes
  • LED displays for speed and time
The OHAUS Digital Microplate Vortex Mixer is ideal for applications that demand repeatable results. Mixer features touchpad controls and LED displays for accurate speed (rpm) and time (minutes and seconds) results. Microprocessor control maintains set speed for strong, consistent mixing action. Timer will display elapsed time or, when programmed to a user defined time limit, the unit will shut off when time reaches zero. Choose from two modes of operation- "Touch" mode which activates mixing when cup head is depressed, or "On" mode when using the microplate attachments for continuous operation.
Operating Features: Microprocessor Control– The variable speed microprocessor control provides consistent uniform mixing action. LED Display-Touch pad controls with easy-to-read, independent LED displays for speed and time allow operator to view both settings at once. Provides repeatable and accurate results every time and is easily visible across lab benches. Timer will display elapsed time or, when programmed to user defined limit, will shut off unit when time reaches zero. Display will show last used settings, even after power has been turned off.
Operating Conditions: Unit can be run in conditions from 4 to 40 °C (39 to 104 °F), 20 to 85% relative humidity, non-condensing.
Ordering information: Mixer includes a cup head, single microplate holder, and a detachable, 3-wire cord and specified plug. Accessories that can be used on the Microplate Vortex Mixer are cup head, single microplate holder and double microplate holder.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty