Would you like to log in again?
Multi Purpose Water Bath Clamps, Ohaus
Supplier: Ohaus
30392240 30392241
470235-398EA 67.25 USD InStock
470235-398 470235-396
Multi Purpose Water Bath Clamps, Ohaus
Clamps General Purpose Clamps
Lab Essentials with Prong Style, Grip Size and Material Options.
  • Nickel-plated zinc construction
  • Additional holder is required in order to attach clamps to lab-frames and other apparatus
Holds a variety of apparatus, including glass tubes and thermometers onto glass water bath walls. Built-in holder grips walls of varying thickness, up to 9 mm. Knurled thumb screw tightens jaws to hold objects firmly. Large water bath clamp is supplied with non-slip vinyl sleeves and, for temperatures above 100 °C, fiberglass covers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty