Would you like to log in again?
Gowns, Cellucap-Melco
Supplier: Cellucap
434TKC 434TCX 434T
77776-604CS 292.44 USD InStock
77776-604 77776-608 77776-606
Gowns, Cellucap-Melco
Disposable Garments
Gowns are designed for fluid-proof protection.
Made of polycoated polypropylene and it covers ties, the neck, and waist.
Packaging: 50 per case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty