Would you like to log in again?
Sklar Edge™ Tungsten Carbide Z-Type Parametrium Scissors, OR Grade, Sklar®
Supplier: Sklar
Sklar Edge™
16-4013 16-4010 16-4008 16-4001 16-4000 16-4014 16-4012 16-4002 16-4006 16-4005 16-4009 16-4004
76192-176EA 1168.02 USD InStock
76192-176 76192-166 76192-174 76192-164 76192-172 76192-162 76192-170 76192-160 76192-158 76192-156 76192-178 76192-168
Sklar Edge™ Tungsten Carbide Z-Type Parametrium Scissors, OR Grade, Sklar®
Scissors
Sklar Edge™ Tungsten Carbide Z-Type Parametrium Scissors are a non-ratcheted finger ring OB/GYN instrument used for cutting Parametrium tissue, also known as cervical stroma, in vaginal hysterectomy procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Z-Type Parametrium Scissors are available in multiple lengths with various curvatures so as to compliment the size and curvature of Z-Type Hysterectomy Forceps. This design allows the two instruments to be used in conjunction without blocking the view of the physician. Sklar Edge™ Tungsten Carbide Scissors provide an exceptionally durable and hard cutting edge. These scissors are designed to stay sharp for a significantly longer period of time than standard scissors. Sklar permanently bonds tungsten carbide inserts to the blades, which maximizes control and precision. Sklar Edge™ scissors can be identified by two gold coated finger rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty