Would you like to log in again?
Lab-Elite™ Certified Reference Materials (CRM), KWIK-STIK, Microbiologics
Supplier: Microbiologics
KWIK-STIK™ Lab-Elite™
0317-CRM 0890-CRM 0687-CRM 0121-CRM 0968-CRM 0345-CRM 0111-CRM 0789-CRM 0441-CRM 0805-CRM 0356-CRM 0902-CRM 0817-CRM 0727-CRM 0262-CRM 0421-CRM 0947-CRM 0261-CRM 01045-CRM 01056-CRM 0697-CRM 0604-CRM
76199-936PK 97.17 USD InStock
76199-936 76200-018 76200-028 76200-204 76200-138 76200-006 76200-008 76200-024 76200-142 76200-032 76200-020 76200-130 76200-012 76199-952 76199-954 76199-942 76200-140 76199-956 76199-934 76200-030 76200-194 76199-944
Lab-Elite™ Certified Reference Materials (CRM), KWIK-STIK, Microbiologics
Control reagents
Our Lab-Elite CRM is designed to help those labs meet the requirements of ISO 17025 standards. They are just one passage from the reference culture and are delivered in our signature KWIK-STIK format.
  • Traceable to reference culture
  • Refrigerated storage is easy and economical
  • Ready-to-use format to save you time and money
  • Strains are one passage from reference culture
  • Technical Support experts available for guidance
This standard specifies that "reference materials shall, where possible, be traceable to SI units of measurements, or to Certified Reference Materials". As a Certified Reference Material, Lab-Elite comes with a comprehensive Certificate of Analysis detailing the strains identity, characteristics and standard deviation. Qualitative QC microorganisms one passage from the reference strains for ultimate authenticity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty