Would you like to log in again?
Martin Tissue Forceps, OR Grade, Sklar®
Supplier: Sklar
19-2166 19-2168 19-2167
76192-408EA 118.17 USD InStock
76192-408 76192-406 76192-410
Martin Tissue Forceps, OR Grade, Sklar®
Forceps
Sklar® Martin Tissue Forceps are thumb forceps used to hold medium weight tissue.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar® Martin Tissue Forceps feature 7x8 horizontal teeth. They are commonly used in OB/GYN procedures. They may also be known as Martin-Nelson Forceps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty