Would you like to log in again?
HiScreen IMAC FF Columns
Supplier: Cytiva
HiScreen™
28-9505-17 28950517
97067-826EA 258.31 USD InStock
97067-826 CA97067-826
HiScreen IMAC FF Columns
Chromatography Columns
HiScreen™ IMAC FF columns for optimising purification of histidine-tagged proteins by Immobilised Metal ion Affinity Chromatography (IMAC). The columns are prepacked with IMAC Sepharose™ 6 fast flow.
  • Conveniently charge with metal ion of choice (Cu²⁺, Zn²⁺, Ni²⁺ or Co²⁺)
  • Prepacked HiScreen™ columns for method optimization and process development
  • High binding capacity
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity
Flow velocity of <600 cm/h; the pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty