Would you like to log in again?
Adson Aneurysm Needle, OR Grade, Sklar®
Supplier: Sklar
26-1830
76193-018EA 78.73 USD InStock
76193-018
Adson Aneurysm Needle, OR Grade, Sklar®
Dissection Needles
The Sklar® Adson Aneurysm Needle is commonly used in intracranial procedures for ligation of arteries and treatment of an aneurysm.
This product has a short, right angled tip with an eye, through which the ligatures are passed. Aneurysm needles are also used for passing ligatures around deep wounds or tubular structures in OB/GYN procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty