Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sklarlite™ Sklarhone™ Metzenbaum-Nelson Bariatric Scissors, OR Grade, Sklar®
Supplier: Sklar
15-9119
76192-028EA 319.45 USD InStock
76192-028
Sklarlite™ Sklarhone™ Metzenbaum-Nelson Bariatric Scissors, OR Grade, Sklar®
Scissors
Sklarlite™ Sklarhone™ Metzenbaum-Nelson Bariatric Scissors are long dissecting scissors most commonly used in Bariatric Surgeries such as gastric bypass, gastric banding, and sleeve gastrectomy. They are also used in Nephrectomy, Cystectomy, and Prostatectomy procedures to dissect and cut tissue.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Metzenbaum-Nelson scissors tend to be longer than standard Metzenbaum scissors, making them ideal for use in thoracic procedures as well. These scissors are not recommended for cutting sutures or drains. The Sklarlite™ Sklarhone™ combination provides the surgeon with a lightweight, precision instrument to make the sharp incisions of a knife with the cutting ability of scissors, all while having greater control and experiencing less fatigue. This instrument is identified by dual black finger rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty