Would you like to log in again?
Senning Needle Holder, OR Grade, Sklar®
Supplier: Sklar
20-2309
76192-508EA 497.39 USD InStock
76192-508
Senning Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Sennning Needle Holder is commonly used in vascular and plastic surgery procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Sklar® Sennning Needle Holder is a ratcheted, finger ring instrument with short jaws and shanks that bend inward slightly, right above the ratchet, similar to those of a Sarot needle holder. This slight bend creates a longer, narrow section of shanks providing access to tight surgical areas.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty