Would you like to log in again?
Mathieu Delicate Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8109 21-8106
76192-704EA 297.33 USD InStock
76192-704 76192-762
Mathieu Delicate Needle Holder, OR Grade, Sklar®
Forceps
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Sklar® Mathieu Needle Holder is a single spring instrument with a plier handle grip that curves inward at the bottom and features a ratchet locking mechanism at the base of the handles that can be easily squeezed to be locked and unlocked. This needle holder is used when more refined suturing is required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty