Would you like to log in again?
TC Mathieu-Ryder Vascular Needle Holder, OR Grade, Sklar
Supplier: Sklar
21-8110
76192-764EA 796.41 USD InStock
76192-764
TC Mathieu-Ryder Vascular Needle Holder, OR Grade, Sklar
Forceps
The Sklar® Tungsten Carbide Mathieu-Ryder needle holder is a single spring, plier handle instruent with a locking ratchet.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Mathieu-Ryder needle holder is used for suturing, commonly in plastic surgeries requiring very fine sutures. It has a fine jaw, versus the regular Mathieu. The Mathieu-Ryder jaw tip profile is similar to the Langenbeck-Ryder. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty