Would you like to log in again?
Econo™ Gelpi Retractor, Floor Grade, Sklar®
Supplier: Sklar
Econo™
21-805
76192-714BX 926.17 USD InStock
76192-714
Econo™ Gelpi Retractor, Floor Grade, Sklar®
Retractors
The Econo™ Gelpi Retractor is a ratcheted, finger ring retractor often used in smaller surgical sites.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Once the targeted tissue has been held back, the locking mechanism allows the retractor to remain in place, and does not require someone to hold it during the procedure. The Gelpi retractor has a single sharp prong on each tip that is turned outward, sideways, at a 90° angle. Econo™ instruments are floor-grade, disposable instruments sold in bulk quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty