Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Econo™ Vannas Scissors, Floor Grade, Sklar®
Supplier: Sklar
Econo™
21-910
76192-864BX 595.42 USD InStock
76192-864
Econo™ Vannas Scissors, Floor Grade, Sklar®
Scissors
Econo™ Vannas scissors are double spring thumb scissors most commonly used in ophthalmic procedures such as iridectomy and capsulotomy, but also used in some vascular procedures or to cut 10-0 and 9-0 sutures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The 6mm, delicate blades are available straight, curved, or angled. Econo™ instruments are floor grade, disposable instruments that eliminate the need for cleaning and reprocessing. Because they are disposable, Econo™ instruments also reduce the likelihood of cross-contamination. They are sold in bulk quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty