Would you like to log in again?
Supercut Sklarhone™ Baby-Metzenbaum Dissecting Scissors, OR Grade, Sklar®
Supplier: Sklar
Supercut Sklarhone™
16-3502 16-3501
76192-010EA 187.97 USD InStock
76192-010 76192-008
Supercut Sklarhone™ Baby-Metzenbaum Dissecting Scissors, OR Grade, Sklar®
Scissors
Supercut Sklarhone™ Baby Metzenbaum Dissecting Scissors combine one knife-like razor edge and one micro-serrated scissor edge that can be used for cutting and dissection in small surgical sites.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
These scissors can be used to cut both delicate and heavy tissue because of their unique design. The micro-serrations of the these scissors minimize tearing of tissue, so cuts and edges are smooth and clean. Sklarhone™ instruments are identified by dual black finger rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty