Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sklar Edge™ Tungsten Carbide Goldman-Fox Scissors, OR Grade, Sklar®
Supplier: Sklar
Sklar Edge™
16-3614 16-3615
76192-098EA 568.05 USD InStock
76192-098 76192-140
Sklar Edge™ Tungsten Carbide Goldman-Fox Scissors, OR Grade, Sklar®
Scissors
Sklar Edge™ TC Goldman-Fox scissors are primarily used in dental procedures to trim gingival tissue, but may also be used in plastic surgery or hemorrhoidectomy and other rectal procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
They have beveled blades, one of which is serrated, that taper into sharp, fine tips. The serrations provide a firm grip on tissue and the tapered blades make small, precise cuts in small surgical areas. Sklar Edge™ Tungsten Carbide scissors provide an exceptionally durable and hard cutting edge, designed to stay sharp for a long period of time. Sklar permanently bonds tungsten carbide inserts to the blades of the scissors which maximizes control and precision. Sklar Edge™ scissors can be identified by two gold coated finger rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty