Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sklar Edge™ Tungsten Carbide Ragnell-Kilner Scissors, OR Grade, Sklar®
Supplier: Sklar
Sklar Edge™
16-3616
76192-142EA 287.05 USD InStock
76192-142
Sklar Edge™ Tungsten Carbide Ragnell-Kilner Scissors, OR Grade, Sklar®
Scissors
Tungsten carbide / stainless steel scissors with blunt blades.
  • Reusable
  • Premium OR-Grade
  • Dual Gold handle
For more than 120 years, Sklar has set the standard for surgical instrumentation. With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure. All Sklar products are industry standard.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty