Would you like to log in again?
Dunhill Artery Forceps, OR Grade, Sklar
Supplier: Sklar
17-2250 17-2251
76192-216EA 67.28 USD InStock
76192-216 76192-214
Dunhill Artery Forceps, OR Grade, Sklar
Forceps
Sklar's® Dunhill Artery Forceps are used to grasp vessels, tissue, and sutures during surgical procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
These forceps compress the artery and hold it out of the way, or clamp and seal blood vessels. The instrument is available in straight or curved serrated blades, and it has a ratcheted finger ring handle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty