Would you like to log in again?
Pean Forceps, OR Grade, Sklar®
Supplier: Sklar
17-1755 17-1757
76192-128EA 33.8 USD InStock
76192-128 76192-126
Pean Forceps, OR Grade, Sklar®
Forceps
Sklar® Pean Forceps are used in multiple procedures as a hemostat for small blood vessels.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar® Pean Forceps feature a ratcheted, finger ring handle that provides a secure, locking grip to halt blood flow. The forceps are known for their fine tips, and short, fully serrated jaws. They are available curved or straight.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty