Would you like to log in again?
Adlerkreutz Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
19-1915 19-1960 19-1916 19-1980
76192-318EA 156.61 USD InStock
76192-318 76192-316 76192-302 76192-300
Adlerkreutz Tissue Forceps, OR Grade, Sklar
Forceps
Sklar® Alderkreutz Tissue Forceps are thumb forceps used for gripping and handling tissue.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar® Alderkreutz Tissue Forceps are available in two lengths, 6" or 8", and feature 2x3 or 4x5 teeth in addition to partially, horizontally serrated tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty