Would you like to log in again?
Stille Barraya Forceps, OR Grade, Sklar®
Supplier: Sklar
19-1990
76192-320EA 100.36 USD InStock
76192-320
Stille Barraya Forceps, OR Grade, Sklar®
Forceps
Sklar® Stille-Barraya Forceps are long thumb style forceps commonly used for grasping organs or dissecting tissue in the stomach and intestines.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty