Would you like to log in again?
Reill Micro Needle Holder with Pen-Type Handle, OR Grade, Sklar®
Supplier: Sklar
20-1552 20-1553 20-1550 20-1551
76192-496EA 1038.28 USD InStock
76192-496 76192-494 76192-492 76192-498
Reill Micro Needle Holder with Pen-Type Handle, OR Grade, Sklar®
Forceps
Sklar's® Reill Micro Needle Holder is most commonly used to hold fine suture needles.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar's® Reill Micro Needle Holder is most commonly used to hold fine suture needles in very small sites or microsurgical procedures. It is available straight or curved, in various lengths, and with or without a catch. The handle has semi-cylandrical (Pen Type), ergonomicly designed, texturized grips for optimal comfort and utilization.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty