Would you like to log in again?
Kilner Needle Holder, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
20-2640 20-2641
76192-518EA 185.51 USD InStock
76192-518 76192-516
Kilner Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Kilner Needle Holder is designed for use with delicate suturing needles for closing wounds.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Kilner Needle Holder is often used in plastic surgery procedures. Like similar instruments, it has a serrated tip for gripping. Unlike the Crile or the Mayo, this product is "S" curved for specific procedures.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty