Would you like to log in again?
Baum-Metzenbaum Tonsil Needle Holder, OR Grade, Sklar®
Supplier: Sklar
20-2725
76192-520EA 109.03 USD InStock
76192-520
Baum-Metzenbaum Tonsil Needle Holder, OR Grade, Sklar®
Forceps
The Baum-Metzenbaum needle holder is an ENT instrument used for suturing during tonsil procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Baum-Metzenbaum needle holder is a ratcheted finger ring ENT instrument used for suturing during tonsil procedures. The curved fenestrated jaw with cross serations ensures a firm grip on the needle. It is similar to the Finochietto and Collier styles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty