Would you like to log in again?
Iodine HPLC Assay, Eagle Biosciences
Supplier: Eagle Biosciences
IOD34-H100
102993-526EA 935 USD InStock
102993-526
Iodine HPLC Assay, Eagle Biosciences
Assays Immunological Assays
The Eagle Biosciences Iodine HPLC Assay kit is intended to determine iodine in urine.
  • Incubation Time: 5 minutes
  • Sample Size: 3 ml
  • Sample Type: Urine
The measurement of free iodine in urine is a valuable tool for the determination of the supply with iodine. The measurement of the direct intake of iodine is rather difficult. Therefore the determination of iodine in urine is well accepted as an index of iodine uptake. The fecal excretion is negligible.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty