Would you like to log in again?
Malondialdehyde (MDA) HPLC Assay, Eagle Biosciences
Supplier: Eagle Biosciences
MDA31-H100
102994-780EA 1235.77 USD InStock
102994-780
Malondialdehyde (MDA) HPLC Assay, Eagle Biosciences
Assays Immunological Assays
The Malondialdehyde (MDA) HPLC Assay kit is intended to determine malondialdehyde in plasma,serum.
  • Incubation Time: 1 hour
  • Sample Size: 20 µL
  • Sample Type: Serum, Plasma
In a healthy body, oxidative and reductive processes are in a balance. Free radicals (reactive oxygen species) are eliminated by antioxidants. In case of a lack of antioxidants free radicals react with cell structures. A reaction of free radicals with unsaturated fatty acids leads to lipid peroxidation products. A reaction of free radicals with poly-unsaturated fatty acids generates malondialdehyde and/or 4-hydroxynonenal. These secondary lipid peroxidation products might react with other molecules in the cell. Modifications of the DNA-based adenine or guanine result in incorrect transcripts. A reaction of free radicals with proteins leads to an alteration or loss of function. The creation of neoAntigenss is possible. NeoAntigenss are recognized by the immune system, thus resulting in autoimmune diseases. The participation of lipid peroxidation products have been discussed in several diseases such as atherosclerosis, tumor genesis, rheumatism and reperfusion injury after transplantation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty