Would you like to log in again?
Vitamin B1 (Thiamin) HPLC Assay, Eagle Biosciences
Supplier: Eagle Biosciences
VB131-H100
102995-086EA 856.54 USD InStock
102995-086
Vitamin B1 (Thiamin) HPLC Assay, Eagle Biosciences
Assays Immunological Assays
The Vitamin B1 (Thiamin) HPLC Assay Kit detects vitamin B1 in EDTA-blood.
  • Incubation Time: 20 minutes
  • Sample Size: 50 µl
  • Sample Type: EDTA Whole Blood
The Eagle Biosciences Vitamin B1 (Thiamin) HPLC Assay makes it possible to determine the vitamin in an easy, fast and precise method. The kit includes all reagents in ready to use form for preparation and separation of the samples with exception of the columns (IC2201rp) and the controls (IC2201ko). Thiamin is found free and as mono-, di- and triphosphate. The different forms can be transferred into each other. The metabolic active form is thiamin diphosphate in the body and thiamin triphosphate in the brain. Thiamin diphosphate (TDP) works as thermostable coenzyme in many enzymes of the carbohydrate and amino acid metabolism. In most reactions TDP catalyzes the cleavage of C-C bonds. In the nervous system vitamin B1 is needed for the stimulation of nerve cells.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty