Would you like to log in again?
Vitamin K1 HPLC Assay, Eagle Biosciences
Supplier: Eagle Biosciences
VK131-H100
102995-112EA 1176.92 USD InStock
102995-112
Vitamin K1 HPLC Assay, Eagle Biosciences
Assays Immunological Assays
The Vitamin K1 HPLC Assay Kit is intended to detect vitamin K1 in plasma and serum.
  • Incubation Time: 10 minutes
  • Sample Size: 1 ml
  • Sample Type: Serum, Plasma
Vitamin K works as cofactor of an oxygen dependent carboxylase, which carboxylates specific glutamic acid residues in g-position. Because of that modification, blood clotting factors can be activated by cleavage. In bone metabolism the g-carboxylation enables osteocalcin to bind to hydroxyapatite of the bone matrix. Vitamin K can be stored only for a short time, whereby deficiency occur after a view days. A lack of vitamin K mainly manifests in clotting disorders. Beside this, disorders in bone metabolism caused by a deficient modification of osteocalcin have been reported. The Eagle Biosciences Vitamin K1 HPLC Assay kit makes it possible to determine the vitamin in an easy, fast and precise method. The kit includes all reagents for the preparation and separation of the samples with exception of the columns (IC2400rp) and the controls (IC2400ko).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty