Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Nicotinamide adenine dinucleotid, oxidized form (NAD, oxidised form) in 50mM Tris/HCl + SIRT Substrate

Supplier: Enzo Life Sciences
Synonyms: Nadide Nicotinamide adenine dinucleotide, oxidized NAD+

BML-KI282-0500
89160-362EA 50.2 USD InStock
89160-362
β-Nicotinamide adenine dinucleotid, oxidized form (NAD, oxidised form) in 50mM Tris/HCl + SIRT Substrate
β-Nicotinamide adenine dinucleotide (NAD, oxidized form)
NAD+ (SIRT Substrate)

50mM Nicotinamide adenine dinucleotide (oxidized form) in 50mM Tris/HCl, pH 8.0, containing 137mM sodium chloride, 2.7mM KCl and 1mM MgCl2.


Formula: C₂₁H₂₇N₇O₁₄P₂
MW: 663.43 g/mol
MDL Number: MFCD00036253
CAS Number: 53-84-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty