Would you like to log in again?
Calbiochem® Human Albumin
Supplier: EMD Millipore
Calbiochem®
12666-10GM 12666-10GM 12666-1GM 12666-100GM 12666-1GM
80055-730EA 90.18 USD InStock
80055-730 80055-728 CA80055-728 80055-726 CA80055-730
Calbiochem® Human Albumin
Proteins and Peptides
This albumin is prepared under non-denaturing conditions by a modification of the Cohn procedure.
  • Contaminants: Ash ≤5.0%; moisture ≤6%; heavy metals ≤20 ppm
  • Heavy metals: ≤20 ppm
  • Toxicity: Standard Handling
Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty