Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sepharose™ Protein A/G

Supplier: Rockland Immunochemicals
Sepharose™
PAG50-00-0002 PAG50-00-0002
75931-792EA 122 USD InStock
75931-792 CA75986-508
Sepharose™ Protein A/G
Proteins and Peptides
Sepharose™ protein A/G is a suspension of sepharose beads containing binding domains from both protein A (from Staphylococcus aureaus) and protein G (from Streptococcus sp.) which is used for immunoprecipitation and purification of monoclonal antibodies.

  • Used to pull down antibody:antigen complexes in immunoprecipitation experiments
  • Suitable for use at 20 µl per immunoprecipitation reaction
  • PSC format: agarose conjugate
  • Physical state: suspension of agarose beads
  • Buffer: 20% (v/v) ethanol

When evenly dispersed approximately 2.0 ml of suspension is equal to 0.5 cc of settled beads of Sepharose™ 4 Fast Flow (highly cross linked, 4% agarose derivative). The bead size is approximately 45 to 165 µm when swollen. Each cc of drained beads will bind approximately 20 to 30 mg human IgG. The exclusion limit (Mr) of the beads is 3×10⁷. The maximum linear flow rate is approximately 1300 cm/hr. This product shows long term stability within pH range of 3 to 9 and short term stability within pH range of 2 to 10. Up to 100 reactions.

The product can be used for binding IgG from human, rabbit, goat, mouse and rat. It has strong binding of human, rabbit, cow, sheep, and goat polyclonal antibodies, mouse IgG2a, IgG2b, IgG3, and rat IgG2a. Protein A/G also has moderate affinity for mouse and rat IgG1 and IgG2c. It binds with weak affinity to rat IgG2b.

Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty