Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aspartate Aminotransferase (AST) (Human) ELISA Kit, BioVision

Supplier: BioVision
AB285299-96TEST
76211-040EA 1516.87 USD InStock
76211-040
Aspartate Aminotransferase (AST) (Human) ELISA Kit, BioVision
Assays ELISAs
A Sandwich ELISA kit to the quantify of human AST in serum and other biological fluids.

  • Easy, convenient and time-saving method to measure the level of AST in human serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
  • Detection Range: 0.313 - 20 ng/ml
  • Sensitivity: < 0.188 ng/ml
  • Assay Precision: Intra-Assay: CV < 8%; Inter-Assay: CV < 10%

Biosynthesis of L-glutamate from L-aspartate or L-cysteine. Important regulator of levels of glutamate, the major excitatory neurotransmitter of the vertebrate central nervous system. Acts as a scavenger of glutamate in brain neuroprotection. The aspartate aminotransferase activity is involved in hepatic glucose synthesis during development and in adipocyte glyceroneogenesis. Using L-cysteine as substrate, regulates levels of mercaptopyruvate, an important source of hydrogen sulfide. Mercaptopyruvate is converted into H2S via the action of 3-mercaptopyruvate sulfurtransferase (3MST). Hydrogen sulfide is an important synaptic modulator and neuroprotectant in the brain. BioVision’s AST ELISA kit is a sandwich ELISA assay for the quantitative measurement of human AST in serum, plasma and cell culture supernatants. The density of color is proportional to the amount of human AST captured from the samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty