Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Norepinephrine (NE) ELISA Kit, BioVision

Supplier: BioVision
AB287789-96TEST
76210-984EA 1340.65 USD InStock
76210-984
Norepinephrine (NE) ELISA Kit, BioVision
Assays ELISAs
A competitive ELISA kit for quantify Norepinephrine biological fluids.

  • Easy, convenient and time-saving method to measure the level of Norepinephrine in serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
  • Detection Range: 15.625 to 1000 pg/ml
  • Sensitivity: <9.375 pg/ml
  • Assay Precision: Intra-Assay: CV <8%; Inter-Assay: CV <10%
  • This assay has high sensitivity and excellent specificity for detection of Norepinephrine. No significant cross-reactivity or interference between Norepinephrine and analogues was observed

Norepinephrine (NE) is a precursor of epinephrine that is secreted by the adrenal medulla and is a well-known central and autonomic neurotransmitter. Norepinephrine is the principal transmitter of most postganglionic sympathetic fibers and of the diffuse projection system in the brain arising from the locus ceruleus. Norepinephrine is also found in plants and is used pharmacologically as a sympathomimetic. BioVision’s Norepinephrine ELISA kit is a competitive ELISA assay for the quantitative measurement of Norepinephrine in serum, plasma and cell culture supernatants. The density of color is proportional to the amount of Norepinephrine captured from the samples.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty