Would you like to log in again?
QuickDetect™ Leptin (Human) ELISA Kit, BioVision
Supplier: BioVision
QuickDetect™
AB285244-96TEST
76211-166EA 1043.76 USD InStock
76211-166
QuickDetect™ Leptin (Human) ELISA Kit, BioVision
Assays ELISAs
A rapid ELISA kit for quantify Leptin in human serum, plasma, urine, cell culture samples
  • Easy, convenient and time-saving method to measure the level of PK in human Serum, plasma, urine, cell culture samples, biological fluid.
  • Detection Range: 35 - 2000 pg/ml
  • Sensitivity: < 12 pg/ml
Leptin, the "satiety hormone", is a hormone made by adipose cells that helps to regulate energy balance by inhibiting hunger. Leptin is opposed by the actions of the hormone ghrelin, the "hunger hormone". Both hormones act on receptors in the arcuate nucleus of the hypothalamus to regulate appetite to achieve energy homeostasis. In obesity, a decreased sensitivity to leptin occurs, resulting in an inability to detect satiety despite high energy stores. BioVision’s Leptin ELISA kit is a sandwich ELISA assay for the quantitative measurement of Leptin in human serum, plasma and cell culture supernatants in 90 minutes. The density of color is proportional to the amount of Leptin captured from the samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty