Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

QuickDetect™ Catecholamine (CA) (Rat) ELISA Kit, BioVision

Supplier: BioVision
QuickDetect™
AB287817-96TEST
76211-050EA 1202.75 USD InStock
76211-050
QuickDetect™ Catecholamine (CA) (Rat) ELISA Kit, BioVision
Assays ELISAs
A rapid ELISA kit to quantify Catecholamine in Rat serum, plasma, urine, cell culture samples.

  • Sensitivity: <1.2 pg/ml
  • Cross Reactivity: No significant cross-reactivity or interference between this analyte and its analogues was observed
  • Easy, convenient and time-saving method to measure the level of Catecholamine in rat serum, plasma, urine, cell culture samples, biological fluid
  • Detection Range: 5 to 320 pg/ml

Catecholamine (CA) is a monoamine, an organic compound that has a catechol and a side-chain amine. Catecholamines are derived from the amino acid tyrosine, which is derived from dietary sources as well as synthesis from phenylalanine. Two catecholamines, norepinephrine and dopamine, act as neuromodulators in the central nervous system and as hormones in the blood circulation. The catecholamine norepinephrine is a neuromodulator of the peripheral sympathetic nervous system but is also present in the blood. It causes general physiological changes that prepare the body for physical activity. Some typical effects are increases in heart rate, blood pressure, blood glucose levels, and a general reaction of the sympathetic nervous system. BioVision’s Catecholamine ELISA kit is a sandwich ELISA assay for the quantitative measurement of CA in rat serum, plasma and cell culture supernatants in 90 minutes. The density of color is proportional to the amount of Catecholamine captured from the samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty