Would you like to log in again?
Express Plasmid Maxiprep Kit (25 min), BioVision
Supplier: BioVision
AB288092-25 AB288092-10TEST AB288092-2TEST
76211-770EA 620.52 USD InStock
76211-770 76211-772 76211-774
Express Plasmid Maxiprep Kit (25 min), BioVision
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
For fast and efficient purification of plasmid DNA from 100-200 mL of E. coli culture.
For fast and efficient purification of plasmid DNA from 100-200 mL of E. coli culture.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty