Would you like to log in again?
PicoProbe™ D-2-Hydroxyglutarate Dehydrogenase Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB239706-100TEST
76211-364EA 980.59 USD InStock
76211-364
PicoProbe™ D-2-Hydroxyglutarate Dehydrogenase Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
Rapid, sensitive & simple assay to measures D-2Hydroxyglutarate Dehydrogenase activity
  • Rapid, sensitive and convenient The kit measures total Activity of D-2-Hydroxyglutatare Dehydrogenase Activity with limit of quantification 4 µU/µl using recombinant D2HGDH
D-2-Hydroxyglutarate Dehydrogenase (D2HGDH: EC 1.1.99.2) is a mitochondrial enzyme which catalyzes the conversion of D-2-Hydroxyglutarate into 2-Oxoglutarate. In humans, mutation of D2HGDH gene could reduce D2HGDH activity which will cause the D-2-Hydroxyglutarate accumulation in body fluids. Increased D-2-Hydroxyglutarate level in body fluids leads to D-2-Hydroxyglutaric Aciduria, a neurometabolic disorder characterized as developmental delay, epilepsy, hypotonia, and dysmorphic features. BioVision’s PicoProbe™ D-2-Hydroxyglutarate Dehydrogenase Assay kit provides a quick and easy method for monitoring D2HGDH activity in a wide variety of samples. In this assay, D2HGDH converts D-2-Hydroxyglutarate into 2-Oxoglutarate and NADH, which further reduces PicoProbe™ to generate an intense fluorescent product (Ex/Em = 535/587 nm). This kit is simple, sensitive and high-throughput adaptable and can detect as low as 4 µU/µl of D2HGDH activity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty