Would you like to log in again?
TERT (Human) ELISA Kit, BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB285288-96TEST
76212-032EA 1516.87 USD InStock
76212-032
TERT (Human) ELISA Kit, BioVision
Assays ELISAs
An ELISA kit to quantify human TERT in human serum, plasma and other biological fluids
  • Easy, convenient and time-saving method to measure the level of TERT in serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
  • Detection Range: 0.156 -10 ng/ml
  • Sensitivity: < 0.094 ng/ml
Telomerase is a ribonucleoprotein enzyme essential for the replication of chromosome termini in most eukaryotes. Active in progenitor and cancer cells. Inactive, or very low activity, in normal somatic cells. Catalytic component of the teleromerase holoenzyme complex whose main activity is the elongation of telomeres by acting as a reverse transcriptase that adds simple sequence repeats to chromosome ends by copying a template sequence within the RNA component of the enzyme. BioVision’s Telomerase reverse transcriptase (TERT) ELISA kit is a sandwich ELISA assay for the quantitative measurement of human TERT in serum, plasma and cell culture supernatants. The density of color is proportional to the amount of TERT captured from the samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty