Would you like to log in again?
Estrogen (Human) ELISA Kit, BioVision
Supplier: BioVision
AB285239-96TEST
76211-642EA 1310.03 USD InStock
76211-642
Estrogen (Human) ELISA Kit, BioVision
Assays ELISAs
An competitive ELISA kit to quantify Estrogen in human serum, plasma and other biological fluids
  • Easy, convenient and time-saving method to measure the level of Estrogen in human serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
  • Detection Range: 15.6-1000 pg/ml
  • Sensitivity: < 9.375 pg/ml
Estrogen is the primary female sex hormone. It is responsible for the development and regulation of the female reproductive system and secondary sex characteristics. Estrogen may also refer to any substance, natural or synthetic, that mimics the effects of the natural hormone. The steroid 17β-estradiol is the most potent and prevalent endogenous estrogen, although several metabolites of estradiol also have estrogenic hormonal activity. Synthetic estrogens are used as part of some oral contraceptives, in estrogen replacement therapy for postmenopausal women, and in hormone replacement therapy for trans women. BioVision’s Estrogen ELISA kit is a Competitive ELISA assay for the quantitative measurement of human Estrogen in serum, plasma and cell culture supernatants. The density of color is proportional to the amount of human Estrogen captured from the samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty