Would you like to log in again?
Estrogen Receptor beta (ERβ) (Human) ELISA Kit, BioVision
Supplier: BioVision
AB285292-96TEST
76211-650EA 1516.87 USD InStock
76211-650
Estrogen Receptor beta (ERβ) (Human) ELISA Kit, BioVision
Assays ELISAs
An Sandwich ELISA kit to quantify human ER-beat in human serum, plasma and other biological fluids
  • Easy, convenient and time-saving method to measure the level of ER-beta in human serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
  • Detection Range: 0.313-20 ng/ml
  • Sensitivity: < 0.188 ng/ml
Nuclear hormone receptor. Binds estrogens with an affinity similar to that of ESR1, and activates expression of reporter genes containing estrogen response elements (ERE) in an estrogen-dependent manner. Isoform beta-cx lacks ligand binding ability and has no or only very low ere binding activity resulting in the loss of ligand-dependent transactivation ability. DNA-binding by ESR1 and ESR2 is rapidly lost at 37 degrees Celsius in the absence of ligand while in the presence of 17 beta-estradiol and 4-hydroxy-tamoxifen loss in DNA-binding at elevated temperature is more gradual. BioVision’s ER-beta ELISA kit is a sandwich ELISA assay for the quantitative measurement of human ER-beta in serum, plasma and cell culture supernatants. The density of color is proportional to the amount of human ER-betacaptured from the samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty