Would you like to log in again?
QuickDetect™ 24(S)-Hydroxycholesterol (Human) ELISA Kit, BioVision, Inc.
Supplier: BioVision
QuickDetect™
AB285293-96TEST
76212-126EA 1746.66 USD InStock
76212-126
QuickDetect™ 24(S)-Hydroxycholesterol (Human) ELISA Kit, BioVision, Inc.
Assays ELISAs
A rapid sandwich ELISA kit to quantify CD8 in human serum, plasma, cell culture supernatnat
  • Easy, convenient and time-saving method to measure the level of 24-OHC in human serum, plasma, culture supernatants and other biological fluids.
  • The entire assay can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results.
  • Detection Range: 0.5 -100 ng/ml
An important cholesterol metabolite (cholesterol 24-hydroxylase) found in the brain. It is an endogenous agonist of the nuclear receptor LXR. It has been shown to induce cell death in neuroblastoma cells, possibly via a necroptosis pathway. 24(S)-Hydroxycholesterol has also been found to be a high affinity ligand for RORα and RORγ (Ki = 25 nM). 24(S)-Hydroxycholesterol levels are elevated in the cerebral spinal fluids of patients with neurogenerative diseases suggesting possible clinical applications. It was also able to reduce Aβ production and increase ER-resident immature amyloid precursor protein (APP) levels in human neuroblastoma SH-Sy5Y cells and CHO cells via inhibition of intracellular amyloid precursor protein trafficking. BioVision’s QuickDetect™ 24-OHC ELISA kit is a sandwich ELISA assay for measuring the quantity of human 24-OHC in serum, plasma and cell culture and other biological fluid. The entire process can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty