Would you like to log in again?
Carbonic Anhydrase (CA) Inhibitor Screening Kit (Colorimetric), BioVision, Inc.
Supplier: BioVision
AB283387-100TEST
76211-840EA 967.01 USD InStock
76211-840
Carbonic Anhydrase (CA) Inhibitor Screening Kit (Colorimetric), BioVision, Inc.
Assays Enzyme Assays
It is simple and can be used to identify and characterize CA inhibitors in a high-throughput format.
  • Simple, highly sensitive, high-throughput compatible
  • Rapid screening of Carbonic Anhydrase ligands
  • Kit includes Acetazolamide, a CA inhibitor, and a stable, recombinant human CA
Carbonic anhydrases (CA; EC: 4.2.1.1) are zinc enzymes present in both prokaryotes and eukaryotes. They efficiently catalyze the reversible hydration of CO2 to bicarbonate. Their important patho-physiological roles in respiration, pH and CO2 homeostasis, secretion, gluconeogenesis, ureagenesis etc. make it an important drug target. CA inhibition in different organs can result in different effects. Inhibition of CA helps in different disease treatment procedures; for example, it can help in the treatment of Glaucoma, urine alkalization, managing epilepsy, sleep apnea, acute mountain sickness etc. BioVision’s CA Inhibitor Screening Kit can be used to screen for potent inhibitors of CA activity. It utilizes the esterase activity of an active CA on an ester substrate which releases a chromogenic product. The released product can be easily quantified using an absorbance microplate reader. In the presence of a CA specific inhibitor, the enzyme loses its activity which results in decrease of absorbance. This assay kit is simple and can be used to identify and characterize CA inhibitors in a high-throughput format. Here, we have used the specific inhibitor Acetazolamide, a potent inhibitor of CA I, CA II, CA IV, CA IX, CA XII etc. except CA III
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty