Would you like to log in again?
Deubiquitinase Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB241002-100TEST
76211-448EA 1017.25 USD InStock
76211-448
Deubiquitinase Activity Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
Straight-forward and Highly Sensitive Assay
  • Simple procedure; takes only ~40 min
  • Fast and convenient
Cell activity and viability is tightly regulated by controlling the production and degradation of the thousands of different proteins in the cell. The proteasome is responsible for the majority of cellular protein degradation; however, drugs targeting the proteasome can have side effects caused by the lack of specificity associated with inhibiting the proteasome itself. Altering the ubiquitination state of target proteins is thus appealing as an alternative approach. Modification of the ubiquitin-mediated proteasome pathway has been shown to be a valid mechanism for treating a variety of diseases, all of which involve dysregulation of cellular proteostasis. As such, it is imperative that these ubiquitination signals also be reversible. The enzymes responsible for cleavage, and hence removal of ubiquitin from ubiquitinated proteins, are known as de-ubiquitinating enzymes, or DUBs. They are proteases that hydrolyze the isopeptide bond between an ubiquitin moiety and a lysine residue on its target protein. By removing the ubiquitin molecule, the protein escapes the fate of proteasomal degradation and remains a viable factor in the cell. BioVision’s Deubiquitinase Activity Assay Kit provides a straight-forward and general measure of deubiquitinase activity by utilizing a fluorescent deubiquitinase substrate to detect activity as low as 0.25 µU with purified enzyme.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty