Would you like to log in again?
15-PGDH Inhibitor Screening Kit (Fluorometric), BioVision, Inc.
Supplier: BioVision
AB283396-100TEST
76211-450EA 1295.49 USD InStock
76211-450
15-PGDH Inhibitor Screening Kit (Fluorometric), BioVision, Inc.
Assays Enzyme Assays
Simple assay to screen/study/characterize COX-2 Inhibitors
  • Simple method to screen 15-PGDH inhibitors
  • High-throughput Compatible
  • Includes 15-PGDH Inhibitor Control,
15-hydroxyprostaglandin dehydrogenase (15-PGDH, EC: 1.1.1.141) catalyzes the oxidation of Prostaglandins (PG) to 15-keto metabolites which show greatly reduced biological activity when compared to Prostaglandins. 15-PGDH catalyzes the rate-limiting step of PG catabolism. More recently, the inhibition of 15-PGDH has been reported as a negative regulator of tissue repair and inhibition of this enzyme can potentiate tissue regeneration in mouse models. Thus, pharmacological inhibition of 15-PGDH could have clinical implications by accelerating potential tissue regeneration after damage. In this assay, 15-PG is oxidized by 15-PDGH generating 15-Keto metabolites and oxidizing NAD. NADH reduces a Fluorometric probe thus, generating an enhanced and stable fluorescence signal (Ex/Em=535/587 nm). In the presence of an inhibitor, the enzymatic activity is inhibited resulting No/Low fluorescence. BioVision’s 15-PGDH Inhibitor Screening Kit offers a rapid, simple, sensitive, and reliable test suitable for high-throughput screening of 15-PGDH inhibitors.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty