Would you like to log in again?
EZCell™ Glutathione Detection Kit (Blue Fluorescence), BioVision, Inc.
Supplier: BioVision
AB284552-100TEST
76211-452EA 651.18 USD InStock
76211-452
EZCell™ Glutathione Detection Kit (Blue Fluorescence), BioVision, Inc.
Assays Cellular Assays
Kit provides fast and easy evaluation of the GSH effectors directly in the living cells.
  • Highly sensitive fluorometric method to measure intra-cellular Glutathione levels in response to a variety of biochemical stimuli
  • Simple & High throughput-adaptable
  • Reproducible, Quantitative tool for screening, studying, and characterizing compounds that affect Glutathione metabolism
Glutathione (GSH) represents major low molecular-weight free thiol in living cells. As the key antioxidant in mammalian cells, intracellular GSH forms rapidly eliminated conjugates with electrophilic xenobiotics, free radicals as well as hydroperoxides. Insufficiency of cellular GSH results in oxidative stress damage and mitochondrial degeneration. Diminished levels of glutathione are symptomatic of cell stress thus; it is advantageous to monitor distribution of the intracellular GSH as an indicator of early apoptosis and cell death. BioVision’s EZCellTM Glutathione Detection Kit (Blue Fluorescence) utilizes Monochlorobimane (MCB), a non-fluorescent and cell-permeable dye, that forms highly fluorescent adducts with GSH (GSH-MCB) detectable at EX/EM= 380/465 ±20 nm respectively. The reaction is catalyzed by glutathione-S-transferase (GST) and the intensity of the fluorescence reflects the amount of intracellular GSH. Hence, our kit provides a useful tool for fast and easy evaluation of the GSH effectors directly in the living cells.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty