Would you like to log in again?
Multidrug Efflux Transporter (MDR1/P-gp) Ligand Screening Kit, BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB284553-100TEST
76211-456EA 612.88 USD InStock
76211-456
Multidrug Efflux Transporter (MDR1/P-gp) Ligand Screening Kit, BioVision
Assays Cellular Assays
The assay is highly sensitive, has a simple no-wash protocol and is high-throughput adaptable.
  • Simple method to screen Pg-p inhibitors
  • High-throughput adaptable
  • Includes Inhibitor Control, Verapamil
P-glycoprotein (P-gp, Multidrug Resistance Protein 1 (MDR1), EC 3.6.3.44) is a member of the ATP-binding cassette (ABC) ATPase superfamily of transmembrane transporter proteins. P-gp has an extremely broad substrate specificity and is capable of transporting a vast array of neutral and anionic lipophilic molecules. P-gp strongly affects the oral absorption, tissue distribution and excretion of many drugs and prevents certain lipophilic drugs from penetrating the blood brain barrier. Overexpression of P-gp confers tumor cells with resistance to chemically and pharmacologically distinct chemotherapeutic drugs (such as doxorubicin, vincristine and paclitaxel) by actively pumping them out of cells. Induction of P-gp expression is a frequent cause of treatment failure and tumor-targeted delivery of P-gp inhibitors is being investigated as a strategy for overcoming chemotherapy resistance. BioVision’s MDR1/P-gp Ligand Screening Kit is designed for rapidly screening test compounds for modulation of efflux transporter activity in MDR1-expressing cell lines. The assay uses a lipophilic non-fluorescent P-gp substrate that readily diffuses through the plasma membrane, where it is hydrolyzed to an active fluorophore by cytosolic esterases. The resulting hydrophilic fluorophore is neither membrane permeable nor a substrate for P-gp, hence it remains trapped inside the cell. In MDR1-expressing cell lines, the lipophilic pro-fluorophore is continuously extruded from the cytosol by P-gp, leading to a low intracellular fluorescence. Inhibition of P-gp-mediated efflux by a test compound leads to increased intracellular fluorescence. Specific transporter activity is quantified by comparison with fluorescence accumulated in the presence and absence of a saturating concentration of the included selective P-gp inhibitor. The assay is highly sensitive, has a simple no-wash protocol and is high-throughput adaptable. The kit contains a complete set of reagents sufficient for performing 100 reactions in a 96-well plate format.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty