Would you like to log in again?
Bacterial Counting Colorimetric Assay Kit, BioVision, Inc.
Supplier: BioVision
AB284554-500TEST AB284554-2500TEST
76211-460EA 536.61 USD InStock
76211-460 76211-462
Bacterial Counting Colorimetric Assay Kit, BioVision, Inc.
Microbiology Test Systems Microorganism Tests
The assay is sensitive and can detect the growth of low density cultures
  • Simple procedure; takes less than ~90 minutes
  • Fast and convenient
The Bacteria Counting Colorimetric Assay Kit is a simple and sensitive assay to quantify bacterial concentration and viability. The assay is based on the cleavage of a water soluble tetrazolium salt (WST) to formazan by mitochondrial dehydrogenases. The color produced is directly proportional to the number of viable bacteria and can be quantified using a microplate reader (Absorbance OD 460 nm). The assay is sensitive and can detect the growth of low density cultures starting with as few as 10 bacteria per well at the time of adding the reagent. The assay can also be used to count the number of living bacteria in a broth culture or to study bacterial viability in response to inhibitors, media types, antibiotics, growth and heterologous protein overexpression conditions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty