Would you like to log in again?
Acidic Mammalian Chitinase Activity Kit (Fluorometric), BioVision
Supplier: BioVision
AB273291-100TEST
76211-466EA 1225.95 USD InStock
76211-466
Acidic Mammalian Chitinase Activity Kit (Fluorometric), BioVision
Assays Enzyme Assays
Simple, specific, ultra-sensitive assay that can detect as low as 0.5 mU/ml of AMCase activity.
  • Simple, rapid & convenient assay to measure Acidic Mammalian Chitinase
  • Includes Positive Control (Acidic Mammalian Chitinase)
Acidic Mammalian Chitinase (AMCase) and Chitotriosidase-1(CHIT1) are the only two mammalian active chitinases that belong to the glycosyl hydrolase-18 family (EC 3.2.1.14). They are chitin-fragmenting hydrolases that hydrolyze the β-(1,4)-linkage between the adjacent N-acetyl glucosamine residues of chitin. In contrast to CHIT1, AMCase exhibits its optimum activity under acidic conditions and is relatively abundant in the gastrointestinal tract and lungs. AMCase is found highly expressed both in animal models used for the study of allergic inflammation and in lungs of human asthma. Recent data suggests a major biological function of AMCase in asthma and the changes in AMCase expression can be used to monitor Th-2/IL-13-mediated immune response and inflammation. BioVision’s Acidic Mammalian Chitinase Activity Assay Kit utilizes the ability of AMCase to cleave a synthetic substrate to release the free fluorophore which can be easily quantified (Ex/Em= 320/445 nm) and it uses a specific Lysis Buffer which distinguishes AMCase activity from CHIT1 activity. This kit provides a simple, specific, ultra-sensitive assay that can detect as low as 0.5 mU/ml of AMCase activity in a variety of biological samples
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty