Would you like to log in again?
Butyrylcholinesterase Activity Kit (Colorimetric), BioVision, Inc.
Supplier: BioVision
AB241010-100TEST
76211-586EA 1259.48 USD InStock
76211-586
Butyrylcholinesterase Activity Kit (Colorimetric), BioVision, Inc.
Assays Enzyme Assays
Simple, rapid & convenient assay to measure Butyrylcholinesterase
  • Simple, rapid & convenient assay to measure Butyrylcholinesterase
  • Includes Positive Control (Butyrylcholinesterase)
Butyrylcholinesterase (EC 3.1.1.8; BChE), also known as plasma cholinesterase or pseudocholinesterase, is a serine hydrolase present in almost all mammalian tissues with the highest levels detected in plasma and liver. BChE hydrolyzes choline esters, as well as other esters and acts as an endogenous scavenger for anticholinesterase agents. BChE in plasma serves as the first line of defense against toxic compounds reaching the bloodstream that might inhibit acetylcholinesterase activity, which is essential in the nervous system. In clinical toxicology and clinical chemistry, determination of BChE activity in plasma is the most commonly used and preferred method to diagnose patients showing symptoms of intoxication. A recent study using genome-wide analysis suggested that BChE is a marker of obesity, insulin resistance and metabolic syndrome, hyperlipidemia, coronary artery disease and hypertension. Serum BChE has been proposed as a marker of low-grade systemic inflammation and a marker of cardiovascular risk factor being even capable to predict mortality. Biovision’s Butyrylcholinesterase Activity Kit is based on the ability of BChE to hydrolyze substrate and produce thiocholine. Thiocholine reacts with 5, 5’-dithiobis (2-nitrobenzoic acid) (DTNB) and generates a yellow chromophore that can be quantified at 412 nm. The assay is simple, sensitive and can detect as low as 0.2 U/ml in variety of samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty