Would you like to log in again?
3D Cell Culture Matrix BME Kit, BioVision, Inc.
Supplier: BioVision
AB241012-100TEST
76211-590EA 1191.17 USD InStock
76211-590
3D Cell Culture Matrix BME Kit, BioVision, Inc.
Assays Cellular Assays
Reproducible, Qualitative tool for screening and characterizing compounds in cell culture studies
  • Reproducible, Qualitative tool for screening, studying, and characterizing compounds in cell culture studies
Three dimensional (3D) cell cultures are artificially-created environments in which cells are permitted to grow or interact with their surroundings in a 3D fashion. 3D cell cultures improve the function, differentiation and viability of cells and recapitulate in vivo microenvironment compared to conventional 2D cell cultures. 3D matrices provide a physiologically relevant screening platform, by mimicking the in vivo responses, for many cell types including cancer and stem cells in developmental morphogenesis, pharmacology, drug metabolism and drug toxicity studies. BioVision offers 3D cell culture matrices, including Basement Membrane Matrix (BME, animal-based), Alginate Hydrogel (plant-based) and proprietary Biovision’s Duo-Matrix Mix, to meet the needs and requirements of various research fields. BioVision’s 3D Culture Matrix Kits provide a standardized, yet user friendly and adaptable to high-throughput strategy for setting up spheroid formations, 3D cell cultures and pharmacological studies.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty